Learn More
DWK Life Sciences KECK™ Clip for Spherical Joints, POM, S
KECK™ spherical ground joints ensure a secure connection.
Brand:  DWK Life Sciences 286340707
 View more versions of this product
Product Code. 15804698
  • 549.00 € / Pack of 100
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
KECK™ spherical ground joint clamps represent a favourable alternative to conventional spherical ground joint clamps made of metal. They are easy to put on and take off. The good spring properties of the POM material prevent scratching of the glass surface. An important contribution to safety in the laboratory. The different colours make the sizes easy to distinguish from each other.
  • Excellent dimensional stability and good spring properties
  • No damage to the glass surface during attachment unlike conventional wire clamps
  • Easy, safe and quick to put on and take off
  • Colour coding per size
  • Cost-effective alternative to conventional spherical ground joint clips made of metal
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100
Securing of Spherical Ground Joints
Dark Blue
POM
Securing of spherical ground joints