missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ KECK™ Clip for Spherical Joints, POM, S

KECK™ spherical ground joints ensure a secure connection.

533.00 € - 595.00 €

Specifications

Quantity 100
Material POM
For Use With (Equipment) Securing of Spherical Ground Joints
For Use With (Application) Securing of spherical ground joints
View More Specs

Products 5
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
15884688
View Documents
DWK Life Sciences™
286340501
Purple
533.00 €
Pack of 100
In Stock
to see stock.
Add to basket
 
15894688
View Documents
DWK Life Sciences™
286340604
Light Blue
549.00 €
Pack of 100
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
15804698
View Documents
DWK Life Sciences™
286340707
Dark Blue
549.00 €
Pack of 100
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
15971581
View Documents
DWK Life Sciences™
286340801
Dark Red
580.00 €
Pack of 100
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
15814698
View Documents
DWK Life Sciences™
286340904
Orange
595.00 €
Pack of 100
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
Description

Description

KECK™ spherical ground joint clamps represent a favourable alternative to conventional spherical ground joint clamps made of metal. They are easy to put on and take off. The good spring properties of the POM material prevent scratching of the glass surface. An important contribution to safety in the laboratory. The different colours make the sizes easy to distinguish from each other.

  • Excellent dimensional stability and good spring properties
  • No damage to the glass surface during attachment unlike conventional wire clamps
  • Easy, safe and quick to put on and take off
  • Colour coding per size
  • Cost-effective alternative to conventional spherical ground joint clips made of metal
Specifications

Specifications

100
POM
Securing of Spherical Ground Joints
Securing of spherical ground joints
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences™ KECK™ Clip for Spherical Joints, POM, S

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.