Learn More
DWK Life Sciences Kimble™ Kontes™ PTFE End Fitting for Chromaflex Columns
Replacement PTFE End Fitting with Bed Support for CHROMAFLEX™ and FLEX-COLUMN™ columns
Brand: DWK Life Sciences 420832-2100
This item is not returnable.
View return policy
Description
Replacement PTFE End Fitting with Bed Support for CHROMAFLEX™ and FLEX-COLUMN™ columns
- PTFE Ending Fitting with Bed Support
- 1.0cm, 2.5cm and 4.8cm options
- Requires attachment with CTFE adapter 420804-0001, not included
Specifications
3.8, 5, 7cm I.D. Chromaflex Column | |
10 mm | |
End Fitting | |
Bed supports |