missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ PTFE End Fitting for Chromaflex Columns
Replacement PTFE End Fitting with Bed Support for CHROMAFLEX™ and FLEX-COLUMN™ columns
136.00 €
Product Code | Brand | Diameter (Metric) Inner | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | For Use With (Equipment) | Price | Quantity & Availability | ||||
15300168
|
DWK Life Sciences
420832-2100 |
10 mm | 3.8, 5, 7cm I.D. Chromaflex Column |
136.00 €
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
||||
Description
Replacement PTFE End Fitting with Bed Support for CHROMAFLEX™ and FLEX-COLUMN™ columns
- PTFE Ending Fitting with Bed Support
- 1.0cm, 2.5cm and 4.8cm options
- Requires attachment with CTFE adapter 420804-0001, not included
Specifications
End Fitting | |
Bed supports |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title