Learn More
DWK Life Sciences Kimble™ Kontes™ Pestles For Dounce All-Glass Tissue Grinders
All-glass component parts.
Brand:  DWK Life Sciences 885302-0002
 View more versions of this product
Product Code. 10389444
  • 107.00 € / Each
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with Dounce (Kontes 885300 series) tissue grinders
  • B (Tight)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.06 oz.
Small clearance pestle, B
0.06 oz., 2mL
2 mL
Dounce tissue grinder