Learn More
DWK Life Sciences Kimble™ Kontes™ Potter-Elvehjem Tissue Grinders
With serrated PTFE pestle and graduated tube
Brand:  DWK Life Sciences 885510-0021
 View more versions of this product
Product Code. 17730888
  • 150.00 € / Each
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
With serrated PTFE pestle and graduated tube
  • Radial serrations on the lower portion of PTFE pestle deliver homogenate into cylindrical portion of tube
  • Stainless-steel pestle shaft
  • Graduations fused into the glass tube
  • Tubes and pestles are interchangeable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
21
6mm
10 mL
1 to 9mL
213mm
128mm
0.23 in.
0.33 oz.
Pestle and graduated tube
8.38 in.
5.03 in.
0.33 oz., 10mL