Learn More
DWK Life Sciences Kimble™ Kontes™ Potter-Elvehjem Tissue Grinders
With serrated PTFE pestle and graduated tube
Brand:  DWK Life Sciences 885510-0022
 View more versions of this product
Product Code. 17740888
  • 203.00 € / Each
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
With serrated PTFE pestle and graduated tube
  • Radial serrations on the lower portion of PTFE pestle deliver homogenate into cylindrical portion of tube
  • Stainless-steel pestle shaft
  • Graduations fused into the glass tube
  • Tubes and pestles are interchangeable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
22
6mm
30 mL
2 to 25mL
225mm
160mm
0.23 in.
1.01 oz.
Pestle and graduated tube
8.85 in.
6.2 in.
1.01 oz., 30mL