Learn More
Thermo Scientific™ KingFisher™ Apex Dx Instrument with 96 Combi Head
The Thermo Scientific KingFisher Apex Dx Purification Instrument enables extraction of nucleic acids, proteins, and cells and has a small, compact footprint, making it ideal for use in diagnostic and development workflows.
Brand: Thermo Scientific™ B5400920
Beschreibung
The Thermo Scientific KingFisher Apex Dx Purification Instrument allows for extraction of nucleic acids, proteins, and cells within a small, compact footprint, making it ideal for use in diagnostic and development workflows. With its intuitive software, automation capabilities, and revolutionary magnetic particle separation technology, this instrument provides high quality, exceptional reliability, and outstanding run-to-run reproducibility for clinicians and assay developers seeking an IVD sample preparation solution for their diagnostic workflow.
The instrument is US IVD and IVDR (EU 2017/746) compliant.
Compact and Stand-alone Instrument
KingFisher Apex Dx is compact and stand-alone, allowing you to easily set up extraction runs, control the instrument, and download run reports from the instrument without the need for a computer.
Adaptable Sample Throughput
Our instrument offers interchangeable 96- and 24-magnet heads and heating blocks, providing you with the flexibility to adjust the sample throughput and volume as needed.
IQOQ Services Designed for Regulatory Compliance
We offer on-site installation qualification and operational qualification (IQ/OQ) services designed to help you comply with regulatory requirements. Instrument setup and IQOQ are included with instrument purchase, performed by Thermo Fisher qualified personnel. OQ is recommended after moving the instrument, upgrading software or hardware, planned maintenance, or critical repairs.
Get started with SmartStart Orientation
SmartStart Orientation is included with instrument purchase, so that you are up and running soon. This orientation, available on-site or remotely, will help you achieve an in-depth understanding of your instrument with engaging and interactive training led by an experienced Field Application Scientist (FAS).
AB Assurance Dx
The AB Assurance Dx service plan is exclusively for IVD instruments and designed to maintain instrument performance to help ensure availability of critical instrument systems in your workflow. The AB Assurance Dx service plan will help keep your laboratory operations running smoothly with planned maintenance, qualification services, proactive instrument monitoring (where available), and—should your instruments require service—a timely response. This service plan is ideal for locations like clinical and IVD laboratories that depend on their IVD instruments and the need to reduce the risk of production downtime. Service and instrument hardware qualification documentation are provided as supporting proof of instrument performance and maintenance history for customer Quality Management Systems (QMS).
Diomni™ Enterprise Software* for Connectivity and Laboratory Ecosystem Management (optional)
Diomni™ Software enables remote browser-based applications, when using with compatible Thermo Fisher Real-Time PCR instruments, such as QuantStudio 7 Pro Dx and QuantStudio 5 Dx, for multiple user and instrument connectivity for the convenience of your laboratory ecosystem management.
Spezifikation
94 Samples | |
79 x 56 x 46 cm | |
Infectious Diseases, Oncology, Diagnostics | |
Case of 1 | |
56 kg |
• KingFisher™ Apex Dx instrument configured with 96 Combi Head, 96 Heating Block, and 96 Deep Well Heating Block |
|
10 in. Touch Display | |
KingFisher™ | |
Room Temperature |
Produktvorschläge
Customers who viewed this item also viewed
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For In Vitro Diagnostic Use.