Learn More
Thermo Scientific™ KingFisher™ Apex Dx Magnetic Heads
The Thermo Scientific KingFisher Apex Dx Purification Instrument allows for extraction of nucleic acids from human specimens with a small, compact footprint making it ideal for use in diagnostic and development workflows.
Brand: Thermo Scientific™ B24079930
Description
KingFisher Apex Dx Magnetic Heads are for use with the KingFisher Apex Dx Purification Instrument. The instrument accommodates two interchangeable magnetic heads (it comes with one included). The interchangeable heads and heating blocks provide flexibility in the extraction of nucleic acids, proteins, and cells with a small, compact footprint, making the instrument ideal for use in diagnostic and development workflows.
With its intuitive software, automation capabilities, and revolutionary magnetic particle separation technology, the KingFisher Apex Dx Purification Instrument provides high quality, exceptional reliability, and outstanding run-to-run reproducibility for clinicians and assay developers seeking an IVD sample preparation solution for their diagnostic workflow.
Specifications
Nucleic Acid Extraction and Cell Separation Instrument | |
Apex Dx 96 Deep Well Head | |
1 head |
Viral Samples | |
Reusable |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
293024010097 | Certificate of Compliance | 28/01/2025 | B24079930 |
293024010088 | Certificate of Compliance | 28/01/2025 | B24079930 |
293024010096 | Certificate of Compliance | 28/01/2025 | B24079930 |
293024010090 | Certificate of Compliance | 28/01/2025 | B24079930 |
293024010087 | Certificate of Compliance | 28/01/2025 | B24079930 |
For in vitro diagnostic use.