missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KingFisher™ Flex Purification System

Prepare samples efficiently from a variety of materials with Thermo Scientific KingFisher Flex Purification Systems, which offer highly versatile, automated magnetic-particle processing for DNA/RNA, protein or cell purification from virtually any source.

Brand:  Thermo Scientific™ 24074420

 View more versions of this product

Product Code. 12579317

  • 4435.00 â‚¬ / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Prepare samples efficiently from a variety of materials with Thermo Scientific KingFisher Flex Purification Systems, which offer highly versatile, automated magnetic-particle processing for DNA/RNA, protein or cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproduciblity and quality.

Find a flexible solution for different types of sample processing needs. With 24 configurations, customers can raise the processing volume up to 5mL, resulting in increased yield of the purified product. For higher throughput needs, 96 samples can be processed with 96 magnet head.

• Fully automated system yields high-speed purification of nucleic acids, proteins and cells.
• High throughput system processes up to 96 samples in less than 20 minutes.
• Expands sample volume by 10-fold with the 24-well format.
• Open and flexible system lets the customer use anymagnetic particle-based kit tomeet the application demands.
• Easy-to-use Thermo Scientific™ BindIt™ Software provides instrument control, protocol creation andmodification.
• Ready-made protocols for different type of applications are available.

Recommended for:
• Genomics and proteomics
• Target identification
• Veterinary assays
• Biomarker discovery
• Quality control
• Forensics

TRUSTED_SUSTAINABILITY
Specifications

Specifications

96 KF PCR head
20 min.
Product Suggestions

Product Suggestions

Videos

Videos

  • Play Video
    Play
    Mute
    Current Time 0:00
    /
    Duration Time 0:00
    Loaded: 0%
    Progress: 0%
    Stream TypeLIVE
    Remaining Time -0:00
     
    Fullscreen
    Share
    Close
    Share Video:

    Thermo Scientific KingFisher Flex Live Demo

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ KingFisher™ Flex Purification System >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Laboratory Use