Learn More
Kimberly-Clark™ Kleenex™ Extra Gel Hand Sanitiser
Fortified with emollients and kills up to 99.99% of a wide range of microorganisms. With a dye and fragrance free formulation, this alcohol gel is dermatologically tested. The Kleenex™ Extra Gel Hand Sanitiser refills are designed for quick and easy installation with a simple push and click.
Brand: Kimberly-Clark™ 6382
102.04 EUR valid until 2025-02-13
Use promo code "24130" to get your promotional price.
Description
- Designed to deliver an unrivalled experience whilst supporting high hand hygiene standards in the workplace.
- This alcohol hand sanitiser kills up to 99.99% of a wide range of microorganisms.
- The hand gel is suitable for use in hygiene-critical areas such as healthcare and food processing environments and meets the requirements of the Biocidal Products Regulation (BPR, Regulation (EU) 528/2012).
- This Kleenex™ hand sanitiser conforms to EN12791, EN13624, EN13727, EN14476, EN14348, EN1500.
- The dye and fragrance free formulation of this gel hand sanitiser is dermatologically tested and is fortified with emollients to support frequent use.
- The hand sanitiser refills are compatible with a range of Aquarius™ hand cleanser dispensers (product codes 6948, 6955, 7124 and 7173).
- The hand sanitiser gel cassettes are designed for quick and easy installation with a simple push and click.
- When the alcohol gel cassette is empty, it can be compressed to conserve space and is fully recyclable once the pump is removed.
Specifications
Sterilizer/Disinfectant Hand Gel | |
Cassette | |
6 x 1 L |
Clear | |
6 L |
Safety and Handling
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.