Learn More
Ecolab Klercide 70/30 IPA Blended with Deionized Water
Klercide 70/30 IPA blended with deionized water is sterile and 0.2 micron filtered.
Brand:  Ecolab 6600956
 View more versions of this product
Product Code. 17835497
  • 1500.00 € / 20 L
Explore more special offers
Description
Description
  • BPR approved
  • Economical, high grade disinfection with easy application
  • 0.2 micron filtered, filled and triple bagged in a Grade C (ISO Class 7) cleanroom
  • Irradiated at no less than 25 kGy to ensure a sterile alcohol
  • Supplied in a trigger spray incorporating the Sterishield Delivery System which ensures in-use sterility for up to 3 months
  • The Sterishield Delivery System is the only fully validated closed system designed to keep the fluid contents of the bottle sterile at all times. This ensures full compliance with GMP Annex 1 requirements that disinfectant solutions are sterile prior to use
  • Certificates of Analysis are available for every batch
  • Full supporting technical documentation is available
Specifications
Specifications
Disinfectant
Neutral
4 x 5 L
Liquid
Sterile
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • 70/30 IPA
Signal Word
  • Danger
Hazard Category
  • Flammable and non-flammable aerosols category 1
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H222-Extremely flammable aerosol.
  • H229-Pressurised container: May burst if heated.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P211-Do not spray on an open flame or other ignition source.
  • P251-Do not pierce or burn, even after use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P410-Protect from sunlight.
  • P412-Do not expose to temperatures exceeding 50°C/122°F.
Supplemental information
  • MIXTURE LIST-Contains : Propan-2-ol