missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-alpha-Dipalmitoyl phosphatidylcholine, 98%, Thermo Scientific Chemicals
97.38 € - 1160.78 €
Chemical Identifiers
CAS | 63-89-8 |
---|---|
Molecular Formula | C40H80NO8P |
Molecular Weight (g/mol) | 734.053 |
MDL Number | MFCD00036903 |
InChI Key | KILNVBDSWZSGLL-KXQOOQHDSA-N |
Synonym | colfosceril palmitate, dppc, 1,2-dipalmitoyl-sn-glycero-3-phosphocholine, 1,2-dipalmitoyl-l-lecithin, dipalmitoylphosphatidylcholine, 1,2-dihexadecanoyl-sn-glycero-3-phosphocholine, colfoscerili palmitas, palmitate de colfosceril, palmitato de colfoscerilo, unii-319x2nfw0a |
PubChem CID | 452110 |
ChEBI | CHEBI:72999 |
IUPAC Name | [(2R)-2,3-di(hexadecanoyloxy)propyl] 2-(trimethylazaniumyl)ethyl phosphate |
SMILES | CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCCCCCCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10626974
|
Thermo Scientific Acros
240571000 |
100 mg |
97.38 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10134670
|
Thermo Scientific Acros
240575000 |
500 mg |
318.59 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10512174
|
Thermo Scientific Acros
240570025 |
2.5 g |
1160.78 €
2.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
63-89-8 | |
734.053 | |
KILNVBDSWZSGLL-KXQOOQHDSA-N | |
452110 | |
[(2R)-2,3-di(hexadecanoyloxy)propyl] 2-(trimethylazaniumyl)ethyl phosphate |
C40H80NO8P | |
MFCD00036903 | |
colfosceril palmitate, dppc, 1,2-dipalmitoyl-sn-glycero-3-phosphocholine, 1,2-dipalmitoyl-l-lecithin, dipalmitoylphosphatidylcholine, 1,2-dihexadecanoyl-sn-glycero-3-phosphocholine, colfoscerili palmitas, palmitate de colfosceril, palmitato de colfoscerilo, unii-319x2nfw0a | |
CHEBI:72999 | |
CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCCCCCCCCC |
Specifications
63-89-8 | |
Authentic | |
Glass bottle | |
MFCD00036903 | |
+6.0° to +8.0° (20°C, 589nm) (c=2, CHCl3) | |
KILNVBDSWZSGLL-KXQOOQHDSA-N | |
[(2R)-2,3-di(hexadecanoyloxy)propyl] 2-(trimethylazaniumyl)ethyl phosphate | |
452110 | |
734.08 | |
Powder |
White | |
98% | |
C40H80NO8P | |
+ 7.00 (20.00°C c=2,CHCl3) | |
colfosceril palmitate, dppc, 1,2-dipalmitoyl-sn-glycero-3-phosphocholine, 1,2-dipalmitoyl-l-lecithin, dipalmitoylphosphatidylcholine, 1,2-dihexadecanoyl-sn-glycero-3-phosphocholine, colfoscerili palmitas, palmitate de colfosceril, palmitato de colfoscerilo, unii-319x2nfw0a | |
CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCCCCCCCCC | |
734.053 | |
CHEBI:72999 | |
≥97.5% (Fatty acid purity) | |
L-α-Dipalmitoyl phosphatidylcholine, 98% |
Safety and Handling
HYGROSCOPIC
Product Identifier
- L-a-Dipalmitoyl phosphatidylcholine
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title