missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ L-Arginine-HCl for SILAC

Study protein expression by mass spectrometry with mass variants of lysine, arginine and other amino acids to perform stable isotope labeling with amino acids in cell culture.

52.25 € - 187.00 €

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Amino Acids
Content And Storage Store at room temperature.
Label or Dye Mass Isotope of Amino Acid
Shipping Condition Room Temperature
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10425725
View Documents
Thermo Scientific™
89989
50 mg
52.25 €
50mg
Please to purchase this item. Need a web account? Register with us today!
12192410
View Documents
Thermo Scientific™
88427
500 mg
187.00 €
500mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thermo Scientific™ Heavy and Light Amino Acids are used to specifically analyze protein expression by mass spectrometry using stable isotope labeling with amino acids in cell culture (SILAC) quantification kits.

SILAC is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of “heavy“ 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization and quantitation of proteins.

Highlights:

  • Efficient – 100% label incorporation into proteins of living cells

  • Flexible – different isotopes of heavy and/or light amino acids for arginine, lysine, leucine and proline enable the quantitation of peptides derived from MS-grade proteases

  • Multiplex capabilities – several alternative isotopes of arginine and lysine are available that allow the analysis of multiple treatment conditions in each experiment

  • High-quality supplements – heavy amino acids with >99% isotope purity

Specifications

Specifications

Mass Spectrometer
Store at room temperature.
Room Temperature
Protein
Protein Labeling
Amino Acids
Mass Isotope of Amino Acid
Cells
Mass Spectrometry
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ L-Arginine-HCl for SILAC

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.