missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ L-Arginine-HCl for SILAC

Study protein expression by mass spectrometry with mass variants of lysine, arginine and other amino acids to perform stable isotope labeling with amino acids in cell culture.

Brand:  Thermo Scientific™ 88427

 View more versions of this product

Product Code. 12192410

  • 187.00 € / 500mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Heavy and Light Amino Acids are used to specifically analyze protein expression by mass spectrometry using stable isotope labeling with amino acids in cell culture (SILAC) quantification kits.

SILAC is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of “heavy“ 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization and quantitation of proteins.

Highlights:

  • Efficient – 100% label incorporation into proteins of living cells

  • Flexible – different isotopes of heavy and/or light amino acids for arginine, lysine, leucine and proline enable the quantitation of peptides derived from MS-grade proteases

  • Multiplex capabilities – several alternative isotopes of arginine and lysine are available that allow the analysis of multiple treatment conditions in each experiment

  • High-quality supplements – heavy amino acids with >99% isotope purity

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mass Spectrometer
Store at room temperature.
Room Temperature
Protein
Protein Labeling
Powder
Amino Acids
Mass Isotope of Amino Acid
Cells
Mass Spectrometry
500 mg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberYL369003 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number88427
Lot NumberYA351961 Certificate TypeCertificate of Analysis Date25/01/2023 Catalog Number88427
Lot NumberXH351960 Certificate TypeCertificate of Analysis Date11/08/2022 Catalog Number88427
Lot NumberXB321628 Certificate TypeCertificate of Analysis Date14/02/2022 Catalog Number88427
Lot NumberWA315080 Certificate TypeCertificate of Analysis Date22/01/2021 Catalog Number88427
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ L-Arginine-HCl for SILAC > L-Arginine-HCl; 500mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.