missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-(+)-Asparagine, 99% Product Code.: 11487004

Thermo Scientific Chemicals L-(+)-Asparagine, 99%

Product Code. 11487004
2500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11487004

Brand: Thermo Scientific Chemicals B21473.0E

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-(+)-Asparagine acts as a precursor to aspartate. It is also used as a diuretic. It is also employed in the biosynthesis of glycoproteins and proteins. It acts as a food additive. It reacts with reducing sugar to yield acrylamide, which finds application in french fries, potato chips, and toasted bread. It plays an important role in the function and development of the brain. Further, it is useful for human body, physically and mentally. In addition to this, it is used in cell culture media and is a component of (2-methoxyethoxy)methyl (MEM) non-essential amino acid solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(+)-Asparagine acts as a precursor to aspartate. It is also used as a diuretic. It is also employed in the biosynthesis of glycoproteins and proteins. It acts as a food additive. It reacts with reducing sugar to yield acrylamide, which finds application in french fries, potato chips, and toasted bread. It plays an important role in the function and development of the brain. Further, it is useful for human body, physically and mentally. In addition to this, it is used in cell culture media and is a component of (2-methoxyethoxy)methyl (MEM) non-essential amino acid solution.

Solubility
Soluble in acids, bases. Insoluble in methanol, ethanol, ether and benzene.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-47-3
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.119
MDL Number MFCD00064401
InChI Key DCXYFEDJOCDNAF-REOHCLBHSA-N
Synonym l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid
PubChem CID 6267
ChEBI CHEBI:17196
IUPAC Name (2S)-2,4-diamino-4-oxobutanoic acid
SMILES C(C(C(=O)O)N)C(=O)N

Specifications

Melting Point 234°C to 236°C (decomposition)
Beilstein 1723527
Merck Index 14,837
Solubility Information Soluble in acids,bases. Insoluble in methanol,ethanol,ether and benzene.
Quantity 2500 g
Formula Weight 132.12
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material L-(+)-Asparagine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ23K034 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberB21473.22,B21473.36,B21473.0E
Lot NumberR22J022 Certificate TypeCertificate of Analysis Date27/06/2023 Catalog NumberB21473.22,B21473,B21473.36,B21473.0E
Lot NumberA0429707 Certificate TypeCertificate of Analysis Date08/11/2022 Catalog NumberB21473.22,B21473.36,B21473.0E
Lot NumberA0437909 Certificate TypeCertificate of Analysis Date03/11/2022 Catalog NumberB21473.22,B21473.36,B21473.0E
Lot NumberU04I011 Certificate TypeCertificate of Analysis Date11/08/2022 Catalog NumberB21473.22,B21473.36,B21473.0E

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-(+)-Asparagine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.