missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-(+)-Asparagine monohydrate, 98+% Product Code.: 11467247

Thermo Scientific Chemicals L-(+)-Asparagine monohydrate, 98+%

Product Code. 11467247
100 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11467247

Brand: Thermo Scientific Chemicals A15012.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-Asparagine is a naturally occurring proteinogenic amino acid, used in biomanufacturing cell culture systems for the production of therapeutic recombinant proteins and monoclonal antibodies. It is used in the biosynthesis of proteins. It is also required for development and function of the brain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Asparagine is a naturally occurring proteinogenic amino acid, used in biomanufacturing cell culture systems for the production of therapeutic recombinant proteins and monoclonal antibodies. It is used in the biosynthesis of proteins. It is also required for development and function of the brain.

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5794-13-8
Molecular Formula C4H10N2O4
Molecular Weight (g/mol) 150.134
MDL Number MFCD00151038
InChI Key RBMGJIZCEWRQES-DKWTVANSSA-N
Synonym l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521Show MoreShow Less
PubChem CID 170358
IUPAC Name (2S)-2,4-diamino-4-oxobutanoic acid;hydrate
SMILES C(C(C(=O)O)N)C(=O)N.O

Specifications

Melting Point ∼245°C (decomposition)
Beilstein 5767869
Merck Index 14,837
Solubility Information Soluble in water.
Density 1.543
Quantity 100 g
Formula Weight 150.14 (132.12 Anhydrous)
Optical Rotation +31° (c=5 in 5N HCl)
Percent Purity ≥98%
Chemical Name or Material L-(+)-Asparagine monohydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10174526 Certificate TypeCertificate of Analysis Date01/05/2023 Catalog NumberA15012.22
Lot NumberT06I050 Certificate TypeCertificate of Analysis Date27/07/2022 Catalog NumberA15012.0E,A15012.36,A15012.22
Lot Number10241809 Certificate TypeCertificate of Analysis Date13/07/2022 Catalog NumberA15012.0E,A15012.36,A15012.22
Lot Number10240726 Certificate TypeCertificate of Analysis Date30/05/2022 Catalog NumberA15012.0E,A15012.36,A15012.22
Lot Number10174526 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15012.0E,A15012.36,A15012.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-(+)-Asparagine monohydrate, 98+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.