Learn More
L-Lysine ethyl ester diisocyanate, 97%, Thermo Scientific Chemicals
Description
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Sparingly soluble in water (0.19 g/L).
Notes
Moisture Sensitive. Store away form water/moisture. Store away from oxidizing agent.

Specifications
Specifications
Color | Yellow to Brown |
Refractive Index | 1.4572 |
Solubility Information | Sparingly soluble in water (0.19g/L). |
Quantity | 5 g |
Formula Weight | 226.23 |
Sensitivity | Moisture sensitive |
Percent Purity | 97% |
Physical Form | Viscous Liquid |
Chemical Name or Material | L-Lysine ethyl ester diisocyanate |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P10L046 | Certificate of Analysis | 12/03/2025 | H64384.06,H64384.14 |
X18K005 | Certificate of Analysis | 25/02/2025 | H64384.06,H64384.14 |
Z26K032 | Certificate of Analysis | 31/12/2024 | H64384.06,H64384.14 |
U05K049 | Certificate of Analysis | 23/08/2024 | H64384.06,H64384.14 |
S06K004 | Certificate of Analysis | 12/06/2024 | H64384.06,H64384.14 |
Safety and Handling


- L-Lysine ethyl ester diisocyanate
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : L-Lysine ethyl esterdiisocyanate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.