missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ L/S™ Digital Process Drive

Digital Process Drives are used to transfer, dispense, and meter in lab, process and washdown environments.

Brand:  Masterflex™ 07575-48

 View more versions of this product

Product Code. 17362660

  • 5720.00 € / 1 unit
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Flow rate 0.006 to 3400 mL/min with L/S tubing—flow rate depends on drive rpm pump head, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating mode— continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication—voltage, current and contact closure
  • Dispense by Volume in mL (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)
  • Antidrip function ensures dispensing accuracy
  • Models with open-head sensor stops the drive when the pump head is opened (when used with sensor-equipped pump head)
  • NEMA 4X (IP66) rated housing for washdown applications
Specifications

Specifications

NEMA 4X (IP66) rated housing for washdown applications
0.006 to 3400 mL/min
50/60
6 ft (1.8 m) Line Cord with IEC 320/CEE 22 Connector Coupler, Watertight Cord Connection, Country-specific Plug/Cord Set
Yes
Non-sterile
Dependent on pump head used
0.1 to 600 rpm
Powder-coated Steel
Graphical LCD
Masterflex™ Tubing
IP66
Steel
Variable
Digital Process Drive
90/260 VAC
Dependent on tubing used
USB, RS232
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.