Learn More
Microspec™ L Shaped Spreaders
Features smooth edges which avoids tearing and digging into agar
Brand:  Microspec™ E1
 View more versions of this product
Product Code. 15732125
Description
Description
  • High Impact Polystyrene construction
  • Full length to avoid contamination
  • Disposable
  • Sterile, aseptically manufactured
  • Ideal for bacterial counts
Specifications
Specifications
Single-use
5.5 in.
Microseal Pouch, Ecopack
High-impact Polystyrene
Aseptically Manufactured
Blue
140 mm
7500
Spreader
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Microspec™ L Shaped Spreaders > No. per Case: 7500