missing translation for 'onlineSavingsMsg'
Learn More
L-Valine methyl ester hydrochloride, 99%, Thermo Scientific Chemicals Product Code.: 11402618

L-Valine methyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 11402618
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11402618

Brand: Thermo Scientific Alfa Aesar A15892.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6306-52-1
Molecular Formula C6H13ClNO2
Molecular Weight (g/mol) 166.63
MDL Number MFCD00012497
InChI Key RFMIEHOYULVMNZ-UHFFFAOYNA-N
Synonym l-valine methyl ester hydrochloride, h-val-ome.hcl, methyl valinate hydrochloride, s-methyl 2-amino-3-methylbutanoate hydrochloride, h-l-val-ome hcl, methyl l-valinate hydrochloride, valine methyl ester hydrochloride, s-2-amino-3-methyl-butyric acid methyl ester hydrochloride, h-val-ome hcl, methyl 2s-2-amino-3-methylbutanoate hydrochlorideShow MoreShow Less
PubChem CID 111190
IUPAC Name methyl (2S)-2-amino-3-methylbutanoate;hydrochloride
SMILES [Cl].COC(=O)C(N)C(C)C

Specifications

Melting Point 166°C (decomposition)
Beilstein 3594960
Quantity 10 g
Formula Weight 167.64
Optical Rotation +16° (c=2 in Water)
Percent Purity 99%
Chemical Name or Material L-Valine methyl ester hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10197900 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15892.09,A15892.18
Lot Number10168399 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15892.09,A15892.18
Lot Number10226663 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15892.09,A15892.18
Lot Number10141614 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15892.09,A15892.18
Lot Number10172201 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15892.09,A15892.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-Valine methyl ester hydrochloride, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.