missing translation for 'onlineSavingsMsg'
Learn More

Elgas™ LA612 Reservoir

Elgas™ LA612 Reservoir is a large volume accessory reservoir suitable for PURELAB™ Option-S 7 and S 15 water purifiers.

Brand:  Elgas™ TAPEZF0040

Product Code. 13125960

  • 2995.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Large volume (40L) reservoir
  • Suitable for PURELAB™ Option-S 7 and S15 water purifiers.
Specifications

Specifications

Option S7/ S15 Water Purifier
Reservoir
40 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Elgas™ LA612 Reservoir > For Use With: Option S7/ S15 Water Purifier

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.