missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Reverse Osmosis Membranes

Choose the required Thermo Scientific™ Reverse Osmosis (RO) Membrane for your Thermo Scientific™ Barnstead™ point of use water purification system, as indicated.

Brand:  Thermo Scientific™ 09.2006

413.27 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 11327141

  • 436.17 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Reverse Osmosis membrane with integrated pretreatment, 6L/hr
Smart2Pure
Reverse Osmosis Membrane (pretreated)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Brochures

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Reverse Osmosis Membranes > Reverse Osmosis membrane with integrated pretreatment, 6L/hr

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.