Learn More
OHAUS™ Lab Lift
OHAUS™ Lab Lift provides secure support for holding glassware at elevated positions.
Brand:  OHAUS™ 30400004
 View more versions of this product
Product Code. 15762419
  • 349.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
  • Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
  • Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lab Lift
Clamp, Support, Lab-Lift, CLR-LBLFTS025
254 x 254 (L x W) Platform
Stainless Steel
89 to 330 mm