Learn More
OHAUS™ Lab Lift
OHAUS™ Lab Lift provides secure support for holding glassware at elevated positions.
161.00 € - 1635.00 €
Product Code | Brand | Material | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Size | Price | Quantity & Availability | ||||
15792419
|
OHAUS™
30400007 |
Aluminum | 102 x 102 (L x W) Platform |
161.00 €
Each |
Estimated Shipment: 18-04-2025
Log in to see stock. |
||||
15702429
|
OHAUS™
30400008 |
Aluminum | 152 x 152 (L x W) Platform |
190.00 €
Each |
Estimated Shipment: 18-04-2025
Log in to see stock. |
||||
15712429
|
OHAUS™
30400009 |
Aluminum | 203 x 203 (L x W) Platform |
226.00 €
Each |
Estimated Shipment: 18-04-2025
Log in to see stock. |
||||
15722429
|
OHAUS™
30400010 |
Aluminum | 254 x 254 (L x W) Platform |
298.00 €
Each |
In Stock
Log in to see stock. |
||||
15722419
|
OHAUS™
30400000 |
Stainless Steel | 76 x 76 (L x W) Platform |
200.00 €
Each |
In Stock
Log in to see stock. |
||||
15732419
|
OHAUS™
30400001 |
Stainless Steel | 102 x 102 (L x W) Platform |
236.00 €
Each |
In Stock
Log in to see stock. |
||||
15742419
|
OHAUS™
30400002 |
Stainless Steel | 152 x 152 (L x W) Platform |
241.00 €
Each |
In Stock
Log in to see stock. |
||||
15752419
|
OHAUS™
30400003 |
Stainless Steel | 203 x 203 (L x W) Platform |
328.00 €
Each |
In Stock
Log in to see stock. |
||||
15762419
|
OHAUS™
30400004 |
Stainless Steel | 254 x 254 (L x W) Platform |
349.00 €
Each |
In Stock
Log in to see stock. |
||||
15772419
|
OHAUS™
30400005 |
Stainless Steel | 305 x 305 (L x W) Platform |
923.00 €
Each |
In Stock
Log in to see stock. |
||||
15782419
|
OHAUS™
30400006 |
Stainless Steel | 406 x 406 (L x W) Platform |
1635.00 €
Each |
In Stock
Log in to see stock. |
||||
Description
- Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
- Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
- Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
Specifications
Lab Lift |