missing translation for 'onlineSavingsMsg'
Learn More

SymLine Lab Sink with Drain

Lab Sink with Drain for integration in fume cupboards or on worktops for easy disposal of liquid solvent waste in large quantities.

Brand:  SymLine 118024

603.20 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 17977250

  • 739.00 € / Each
Estimated Shipment: 04-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made of dissipative PE-HD-EL
  • Base with integrated slope for complete emptying
  • Drain with removable sieve
Specifications

Specifications

Black
Laboratory Sink
400 x 300 x 190 mm
HDPE
Pipe 32 mm (OD), Vertical
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SymLine Lab Sink with Drain > Lab Sink with Drain; Thread: Pipe 32 mm (OD), Vertical

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.