missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Cytokeratin 14 Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Cytokeratin 14 Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-115-B1

 View more versions of this product

Product Code. 12669146

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Keratin 14 belongs to the type A (acidic) subfamily of low molecular weight keratins and exists in combination with keratin 5. Keratin 14 has been studied as a prognostic marker in breast cancer.

Host Species: Mouse

Clone: LL002

Isotype: IgG3

Species Reactivity: Human and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide of 15 amino acid residues from the C-terminus of human Cytokeratin 14

Molecular Weight: 50 kDa

Positive Control: A431 cells; Skin or Squamous cell CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cytokeratin 14 Ab-1
Monoclonal
200μg/mL
Mouse
500 μL
Cytoskeletal Protein
Human, Rat
IgG3
Immunohistochemistry (Paraffin), Western Blot
LL002
Biotin
A synthetic peptide of 15 amino acid residues from the C-terminus of human keratin 14
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.