Learn More
Epredia™ Lab Vision™ Cytokeratin 14 Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide
Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Cytokeratin 14 Ab-1, Mouse Monoclonal Antibody.
124.44 € - 611.00 €
Specifications
Antigen | Cytokeratin 14 Ab-1 |
---|---|
Clone | LL002 |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Host Species | Mouse |
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
12659146
|
Epredia™
MS-115-B0 |
Concentrated | Biotin | 100 μL | N/A | ||||
12669146
|
Epredia™
MS-115-B1 |
Concentrated | Biotin | 500 μL | N/A | ||||
12649146
|
Epredia™
MS-115-B |
Concentrated | Biotin | 1 mL | N/A | ||||
12689146
|
Epredia™
MS-115-P0 |
Concentrated | Unconjugated | 100 μL | N/A | ||||
12609156
|
Epredia™
MS-115-P1ABX |
Concentrated | Unconjugated | 100 μL | N/A | ||||
12629156
|
Epredia™
MS-115-PABX |
Concentrated | Unconjugated | 200 μL | N/A | ||||
12699146
|
Epredia™
MS-115-P1 |
Concentrated | Unconjugated | 500 μL |
323.00 €
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12679146
|
Epredia™
MS-115-P |
Concentrated | Unconjugated | 1 mL |
422.00 €
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12659156
|
Epredia™
MS-115-R7 |
Prediluted | Unconjugated | 7 mL |
212.00 €
7mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Keratin 14 belongs to the type A (acidic) subfamily of low molecular weight keratins and exists in combination with keratin 5. Keratin 14 has been studied as a prognostic marker in breast cancer.Host Species: Mouse
Clone: LL002
Isotype: IgG3
Species Reactivity: Human and Rat. Others not known.
Epitope: C-terminal
Immunogen: A synthetic peptide of 15 amino acid residues from the C-terminus of human Cytokeratin 14
Molecular Weight: 50 kDa
Positive Control: A431 cells; Skin or Squamous cell CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
Cytokeratin 14 Ab-1 | |
Immunohistochemistry (Paraffin), Western Blot | |
Mouse | |
Cytoskeletal Protein | |
A synthetic peptide of 15 amino acid residues from the C-terminus of human keratin 14 | |
Primary |
LL002 | |
Monoclonal | |
RUO | |
Human, Rat | |
IgG3 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.