missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 18 Ab-1 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.

Brand:  Epredia MS-142-P0

 View more versions of this product

Product Code. 12639786

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Keratin 18, which belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with keratin 8. It was reported that tissues from gastrointestinal tract are positive for both keratin 8 and 18 but do not contain keratin 14. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for keratin 18.

Host Species: Mouse

Clone: DC10

Isotype: IgG1

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: Human breast cancer PMC 42 cells

Molecular Weight: 45 kDa

Positive Control: HT29 cells, Skin, Breast or Lung CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
Specifications

Specifications

Cytokeratin 18 Ab-1
Monoclonal
200μg/mL
Mouse
100 μL
Cytoskeletal Protein
Human
IgG1
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
DC10
Unconjugated
Human breast cancer PMC 42 cells
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.