Learn More
Cytokeratin 18 Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.
124.00 € - 529.00 €
Specifications
Antigen | Cytokeratin 18 Ab-1 |
---|---|
Clone | DC10 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Host Species | Mouse |
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
12609786
|
Epredia
MS-142-B0 |
Concentrated | Biotin | 100 μL | N/A | ||||
12619786
|
Epredia
MS-142-B1 |
Concentrated | Biotin | 500 μL | N/A | ||||
12699776
|
Epredia
MS-142-B |
Concentrated | Biotin | 1 mL | N/A | ||||
12639786
|
Epredia
MS-142-P0 |
Concentrated | Unconjugated | 100 μL | N/A | ||||
12659786
|
Epredia
MS-142-P1ABX |
Concentrated | Unconjugated | 100 μL | N/A | ||||
12679786
|
Epredia
MS-142-PABX |
Concentrated | Unconjugated | 200 μL | N/A | ||||
12649786
|
Epredia
MS-142-P1 |
Concentrated | Unconjugated | 500 μL |
248.00 €
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12629786
|
Epredia
MS-142-P |
Concentrated | Unconjugated | 1 mL |
365.00 €
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12609796
|
Epredia
MS-142-R7 |
Prediluted | Unconjugated | 7 mL |
212.00 €
7mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Keratin 18, which belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with keratin 8. It was reported that tissues from gastrointestinal tract are positive for both keratin 8 and 18 but do not contain keratin 14. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for keratin 18.Host Species: Mouse
Clone: DC10
Isotype: IgG1
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: Not determined
Immunogen: Human breast cancer PMC 42 cells
Molecular Weight: 45 kDa
Positive Control: HT29 cells, Skin, Breast or Lung CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Specifications
Cytokeratin 18 Ab-1 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Mouse | |
Cytoskeletal Protein | |
Human breast cancer PMC 42 cells | |
Primary |
DC10 | |
Monoclonal | |
IVD | |
Human | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.