missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Pepsin Solution for Enzyme-Induced Epitope Retrieval

Improve the immunoreactivity of many antigens in formaldehyde fixed tissues with Epredia™ Lab Vision™ Pepsin Solution for Enzyme-Induced Epitope Retrieval.

Brand:  Epredia™ AP-9007-005


Product Code. 12628296

  • 38.00 € / 5mL
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • pH 2.0
  • Single-component, ready-to-use format

Storage Conditions: Store at 4°C. Stable for 12 months when stored at 2° to 8°C.
Specifications

Specifications

5 mL
Pepsin
2
Store at 4°C
Immunohistochemistry
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5 ML PEPSIN SOLUTION FOR ENZYME-INDUCED EPITOPE Retrieval (ready to use) CE IVD 5mL

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : pepsin

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.