missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Perforin Ab-2, Mouse Monoclonal Antibody
Employ one marker in a comprehensive antibody portfolio that addresses all major disease states Epredia™ Perforin Ab-2, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1834-S1
This item is not returnable.
View return policy
Description
Perforin is a potent cytolytic pore-forming protein. It is a specific marker of functionally active cytotoxic T-lymphocytes (CTLs) and natural killer (NK) cells. Perforin is a key effector mechanisms in T cell-mediated cytotoxicity. It mediates cytolysis of target cells by membrane damage and apoptosis.Host Species: Mouse
Clone: 5B10
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: C-terminal
Immunogen: Recombinant protein corresponding to C-terminal region of human perforin
Molecular Weight: 70kDa
Positive Control: Lymphoma
Cellular Localization: Granular and cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Perforin Ab-2 | |
Monoclonal | |
Unconjugated | |
Recombinant protein corresponding to C-terminal region of human perforin | |
IVD | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
5B10 | |
Mouse | |
500 μL | |
T-Cell | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction