Learn More
Epredia™ Lab Vision™ Perforin Ab-2, Mouse Monoclonal Antibody
Employ one marker in a comprehensive antibody portfolio that addresses all major disease states Epredia™ Perforin Ab-2, Mouse Monoclonal Antibody.
162.00 € - 769.00 €
Specifications
Antigen | Perforin Ab-2 |
---|---|
Clone | 5B10 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12620247
|
Epredia™
MS-1834-S0 |
Concentrated | 100 μL | N/A | |||||
12640247
|
Epredia™
MS-1834-S1 |
Concentrated | 500 μL |
512.00 €
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12630247
|
Epredia™
MS-1834-S |
Concentrated | 1 mL |
769.00 €
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12610247
|
Epredia™
MS-1834-R7 |
Prediluted | 7 mL |
399.00 €
7mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Perforin is a potent cytolytic pore-forming protein. It is a specific marker of functionally active cytotoxic T-lymphocytes (CTLs) and natural killer (NK) cells. Perforin is a key effector mechanisms in T cell-mediated cytotoxicity. It mediates cytolysis of target cells by membrane damage and apoptosis.Host Species: Mouse
Clone: 5B10
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: C-terminal
Immunogen: Recombinant protein corresponding to C-terminal region of human perforin
Molecular Weight: 70kDa
Positive Control: Lymphoma
Cellular Localization: Granular and cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
1. Lichtenheld M G, et al. (1988) Nature, 335:448-451.
2. Hameed A, et. al. American Journal of Pathology, 1992, 140(5):1025-30.
Specifications
Perforin Ab-2 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Human | |
IgG1 |
5B10 | |
Monoclonal | |
Mouse | |
T-Cell | |
Recombinant protein corresponding to C-terminal region of human perforin | |
Primary |
Safety and Handling
- APD Primary Antibody Products, Antibody Diluent
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P363-Wash contaminated clothing before reuse.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.