Learn More
Labconco™ SpillStopper™ Work Surface for 4 Ft. Protector™Premier™Hoods
Labconco SpillStopper™ Work Surface is dished and contoured to conform to the interior liner to contain spills.
Brand:  Labconco™ 9500410
 View more versions of this product
Product Code. 15916160
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Corrosion-resistant epoxy resins molded
  • Front edge with large radius to aerodynamically direct airflow into hood
  • Pre-drilled holes for venting and plumbing pass-throughs provided
  • Some surfaces with cutouts for surface-mounted service fixture and/or 6 x 3 in. (15.2 x 7.6cm) cup sinkcup sink
  • Flat work surfaces may also be coupled with adjustable height base stand to provide auxiliary workspace
Order Info
Service fixture and cup sink sold separately
Specifications
Specifications
Left Rear
Epoxy Resin