Learn More
Labconco™ SpillStopper™ Work Surface for 4 Ft. Protector™Premier™Hoods
Labconco SpillStopper™ Work Surface is dished and contoured to conform to the interior liner to contain spills.
1510.00 € - 1690.00 €
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
15916160
|
Labconco™
9500410 |
Left Rear |
1690.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16338705
|
Labconco™
9500400 |
No Cutouts |
1510.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Corrosion-resistant epoxy resins molded
- Front edge with large radius to aerodynamically direct airflow into hood
- Pre-drilled holes for venting and plumbing pass-throughs provided
- Some surfaces with cutouts for surface-mounted service fixture and/or 6 x 3 in. (15.2 x 7.6cm) cup sinkcup sink
- Flat work surfaces may also be coupled with adjustable height base stand to provide auxiliary workspace
Order Info
Service fixture and cup sink sold separately
Specifications
Epoxy Resin |