Learn More
Labconco™ Stand-Still™ Isolation Platform
The Labconco Stand-Still™ Isolation Platform is used for supporting microscopes on Purifier™ Cell Logic™+ Biosafety Cabinets with Scope-Ready™ package.
358.00 €
Products 1
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
15538854
View Documents
Labconco™
4028200
12 x 18.75 in.
358.00 €
Each
Estimated Shipment: 14-02-2025
to see stock.
Add to basket
Description
Description
  • Non-porous, autoclavable Type 304 stainless steel base with 1.5 in. (0.4cm) high isolation pads
  • Reduces vertical and horizontal vibrations up to threefold, allowing proper airflow through cabinet
Stand-Still XL Platform
Requires use of Pure-Vu™ XL Seal 4027810 or 4027811 (Part Numbers)