missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Label Maker Cartridge: Nylon Cloth

Label Maker Cartridge: Nylon Cloth (B-499) wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.

Brand:  Brady™ 143356

 View more versions of this product

Product Code. 12719546

  • 74.75 € / Each
Estimated Shipment: 22-11-2024
to see stock.
Edge
Explore more special offers
Add to basket

You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy

Description

Description

Label Maker Cartridge: Nylon Cloth (B-499) wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.

  • Matte white polyamide coated nylon cloth material has a permanent acrylic adhesive and is thermal transfer printable
  • Recommended for indoor use
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted
  • All-in-one cartridge includes ribbon and label supply
Specifications

Specifications

White
Nylon Cloth
12.70 mm
2.54 cm
0.165 mm
1 x 0.5 in.
Non-sterile
Matte White
Rectangle with Rounded Corners
-40°F to +194°F
UL, CSA, DIN VDE 0472 Part 815, ASTM D
1 Ea.
BMP51 Label Printer, BMP53 Label Printer
0.5 in.
1 in.
0.0065 in.
9.53 mm
Permanent
BMP51/53 Labelmaker Labels
9.52 mm dia. Vial top
80 lb./in
-40°C to +90°C
Wiremarking and General Labeling Applications
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers