Learn More
Nickel Electro™ Laboratory Stainless Steel Crucible Lid
Complete a stainless steel crucible with a non-magnetic, bright finish lid. Nickel Electro™ Laboratory Stainless Steel Crucible Lid is flat with a depression for centering and includes an extension on one side that serves as a handle.
Brand:  Nickel Electro™ 163
 View more versions of this product
Product Code. 11489599
  • 67.40 € / Pack of 10
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Includes: DiameterMetricOuter
Requires : Autoclavable,dimMaterial,ProductTypeSpecs,Quantity,Shape
Description
Description
  • Non-magnetic
  • Stamped with the word Stainless Steel to assist in the identification of material when in use
  • Flat shape with a depression for centering on crucibles
  • Includes an extension on one side that serves as a handle
  • Crevice free
Non magnetic, bright finish, crevice free lid, handle
Specifications
Specifications
Stainless Steel
Autoclavable
Crucible lid
10 Pk.
30 mm
Flat