Learn More
Welch™ Laboratory Vacuum System with Glass Ballast Valve
The Laboratory Vacuum System is used to create a controlled vacuum environment, and is commonly used for applications such as filtration, distillation, and solvent evaporation amongst others. The glass ballast valve allows for precise control of the flow of gas into the system.
Brand: Welch™ 115037
Description
- Facilitates the removal of gas or vapor
- Glass ballast valve allows for fine-tuning of the vacuum
- Chemically resistent
- Suction-side separator with round bottom flask
- Assembled, ready for connection

Specifications
2 L/min. (at 60 Hz in m3), 1.8 L/min. (at 50 Hz in m3) | |
230 V 50/60 Hz | |
310 mm | |
<45 dB | |
15 kg | |
50/60 Hz | |
445 mm | |
360 mm | |
Gas or vapor removal | |
<2 mbar |
Glass Ballast Valve | |
360 x 310 x 445 mm | |
LVS 201 T | |
33.1 lb. | |
33 L/min. | |
17.5 in. | |
14.2 in. | |
Vacuum Pump | |
3 | |
230 V |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.