missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Lactic Acid 88% Solution
ACS Reagent Grade
Brand: Melford L17500-4000.0
This item is not returnable.
View return policy
Description
ACS Reagent Grade
ACS Reagent GradeSpecifications
Lactic Acid | |
ACS Reagent Grade | |
4 L |
50-21-5 | |
C3H6O3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction