missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Lactic Acid 88% Solution
ACS Reagent Grade
47.20 € - 199.90 €
Specifications
Chemical Name or Material | Lactic Acid |
---|---|
CAS | 50-21-5 |
Purity Grade Notes | ACS Reagent Grade |
Molecular Formula | C3H6O3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30018371
|
Melford
L17500-500.0 |
500 mL |
47.20 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30018370
|
Melford
L17500-4000.0 |
4 L |
199.90 €
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30018369
|
Melford
L17500-1000.0 |
1 L |
66.20 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACS Reagent Grade
ACS Reagent GradeSpecifications
Lactic Acid | |
50-21-5 | |
ACS Reagent Grade | |
C3H6O3 |