Learn More
Landiolol hydrochloride, MedChemExpress
MedChemExpress Landiolol hydrochloride (ONO1101 hydrochloride) is a highly beta1 selective ultra-short acting beta-blocker (β1/β2 selectivity = 255:1, a half-life of 4 min), acts as an adrenoceptor antagonist.
Brand: Medchem Express HY-100607A
This item is not returnable.
View return policy
Description
Landiolol hydrochloride (ONO1101 hydrochloride) is a highly beta1 selective ultra-short acting beta-blocker (β1/β2 selectivity = 255:1, a half-life of 4 min), acts as an adrenoceptor antagonist.Specifications
Landiolol hydrochloride | |
White | |
C25H40ClN3O8 | |
ONO1101 hydrochloride | |
O=C(OC[C@H]1OC(C)(C)OC1)CCC2=CC=C(OC[C@@H](O)CNCCNC(N3CCOCC3)=O)C=C2.[H]Cl | |
546.05 | |
Research | |
COVID-19-immunoregulation |
144481-98-1 | |
Research | |
5 mg | |
DMSO : 250 mg/mL (457.83 mM; Need ultrasonic) ∣H2O : ≥ 100 mg/mL (183.13 mM) | |
546.05 | |
98.0% | |
Powder |