Learn More
Landiolol hydrochloride, MedChemExpress
MedChemExpress Landiolol hydrochloride (ONO1101 hydrochloride) is a highly beta1 selective ultra-short acting beta-blocker (β1/β2 selectivity = 255:1, a half-life of 4 min), acts as an adrenoceptor antagonist.
POA - 70.20 €
Specifications
Chemical Name or Material | Landiolol hydrochloride |
---|---|
CAS | 144481-98-1 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C25H40ClN3O8 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16483098
|
Medchem Express
HY-100607A |
10 mg |
70.20 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16415837
|
Medchem Express
HY-100607A |
5 mg |
POA
5mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Landiolol hydrochloride (ONO1101 hydrochloride) is a highly beta1 selective ultra-short acting beta-blocker (β1/β2 selectivity = 255:1, a half-life of 4 min), acts as an adrenoceptor antagonist.Specifications
Landiolol hydrochloride | |
White | |
C25H40ClN3O8 | |
DMSO : 250 mg/mL (457.83 mM; Need ultrasonic) ∣H2O : ≥ 100 mg/mL (183.13 mM) | |
546.05 | |
98.0% | |
Powder |
144481-98-1 | |
Research | |
ONO1101 hydrochloride | |
O=C(OC[C@H]1OC(C)(C)OC1)CCC2=CC=C(OC[C@@H](O)CNCCNC(N3CCOCC3)=O)C=C2.[H]Cl | |
546.05 | |
Research | |
COVID-19-immunoregulation |
Safety and Handling
H315∣H319
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.