Learn More
Landiolol hydrochloride, MedChemExpress
MedChemExpress Landiolol hydrochloride (ONO1101 hydrochloride) is a highly beta1 selective ultra-short acting beta-blocker (β1/β2 selectivity = 255:1, a half-life of 4 min), acts as an adrenoceptor antagonist.
Brand:  Medchem Express HY-100607A
Encompass_Preferred
 View more versions of this product
Product Code. 16483098
  • 70.20 € / 10mg
Explore more special offers
Description
Description
Landiolol hydrochloride (ONO1101 hydrochloride) is a highly beta1 selective ultra-short acting beta-blocker (β1/β2 selectivity = 255:1, a half-life of 4 min), acts as an adrenoceptor antagonist.
Specifications
Specifications
Landiolol hydrochloride
White
C25H40ClN3O8
ONO1101 hydrochloride
O=C(OC[C@H]1OC(C)(C)OC1)CCC2=CC=C(OC[C@@H](O)CNCCNC(N3CCOCC3)=O)C=C2.[H]Cl
546.05
Research
COVID-19-immunoregulation
144481-98-1
Research
10 mg
DMSO : 250 mg/mL (457.83 mM; Need ultrasonic) ∣H2O : ≥ 100 mg/mL (183.13 mM)
546.05
98.0%
Powder