missing translation for 'onlineSavingsMsg'
Learn More

3M™ Lane and Safety Marking Tape 471

Strength, conformability, instant adhesion and long-term clean removal make this tape an ideal choice for all but the heaviest floor and safety marking applications. Commonly used for marking hazards and aisles, color-coding, paint masking, decorative trim, and splicing, this tape comes individually wrapped and conveniently packaged.

740.00 € - 765.00 €

Specifications

Adhesive Type Rubber-based Adhesive
Dimensions (L x W) 33 m x 50 mm
Material Vinyl
Packaging Quantity 12 Rolls
Thickness 0.14 mm
View More Specs

Products 6
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
17701699
View Documents
3M™
7100260598
White
765.00 €
12 rolls
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
 
17781679
View Documents
3M™
7100260125
Red
740.00 €
12 rolls
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
 
17741689
View Documents
3M™
7100260283
Yellow
740.00 €
12 rolls
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
 
17711689
View Documents
3M™
7100260162
Green
765.00 €
12 rolls
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
 
17791679
View Documents
3M™
7100260160
Blue
765.00 €
12 rolls
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
 
17701689
View Documents
3M™
7100260161
Black
740.00 €
12 rolls
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
 
Description

Description

  • Maintains color when exposed to abrasion, wear and many solvents
  • Unique stretch properties enable tape to remain stretched to conform to uneven surfaces without lifting and retracting
  • Ideal for use on curved and irregular surfaces
  • Easy to apply, high holding strength on many surfaces
  • Available in many colors (yellow, white, red, black, brown, green, orange, purple, blue and transparent)
  • Clean removal from many surfaces to help reduce clean-up and labor costs
Specifications

Specifications

Rubber-based Adhesive
Vinyl
0.14 mm
33 m x 50 mm
12 Rolls
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ Lane and Safety Marking Tape 471

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.