Learn More
3M™ Lane and Safety Marking Tape 471
Strength, conformability, instant adhesion and long-term clean removal make this tape an ideal choice for all but the heaviest floor and safety marking applications. Commonly used for marking hazards and aisles, color-coding, paint masking, decorative trim, and splicing, this tape comes individually wrapped and conveniently packaged.
Brand:  3M™ 7100260598
 View more versions of this product
Product Code. 17701699
  • 765.00 € / 12 rolls
Estimated Shipment: 11-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Maintains color when exposed to abrasion, wear and many solvents
  • Unique stretch properties enable tape to remain stretched to conform to uneven surfaces without lifting and retracting
  • Ideal for use on curved and irregular surfaces
  • Easy to apply, high holding strength on many surfaces
  • Available in many colors (yellow, white, red, black, brown, green, orange, purple, blue and transparent)
  • Clean removal from many surfaces to help reduce clean-up and labor costs
TRUSTED_SUSTAINABILITY
Specifications
Specifications
33 m x 50 mm
0.14 mm
White
12 Rolls
Rubber-based Adhesive
Vinyl
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M™ Lane and Safety Marking Tape 471 > Lane and Safety Marking Tape 471; Adhesive Type: Rubber-based Adhesive; Color: White; Dimensions (L x W): 33 m x 50 mm; Thickness: 0.14 mm; Material: Vinyl