Learn More
Lanthanum(III) bromide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals
114.00 € - 1325.00 €
Chemical Identifiers
CAS | 13536-79-3 |
---|---|
Molecular Formula | Br3La |
Molecular Weight (g/mol) | 378.62 |
MDL Number | MFCD00016161 |
InChI Key | XKUYOJZZLGFZTC-UHFFFAOYSA-K |
IUPAC Name | lanthanum(3+) tribromide |
SMILES | [Br-].[Br-].[Br-].[La+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11348697
|
Thermo Scientific Alfa Aesar
035800.04 |
2 g |
114.00 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11338697
|
Thermo Scientific Alfa Aesar
035800.09 |
10 g |
381.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11405080
|
Thermo Scientific Alfa Aesar
035800.18 |
50 g |
1325.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a scintillation material in certain application, as a source in the synthesis of lanthanum & its derivatives CsNaLa6Br14N2 and La3Br6N which contain rare bitetrahedral structural units..
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Chemical Identifiers
13536-79-3 | |
378.62 | |
XKUYOJZZLGFZTC-UHFFFAOYSA-K | |
[Br-].[Br-].[Br-].[La+3] |
Br3La | |
MFCD00016161 | |
lanthanum(3+) tribromide |
Specifications
13536-79-3 | |
Br3La | |
Soluble in water. | |
[Br-].[Br-].[Br-].[La+3] | |
378.62 | |
99.9% | |
Hygroscopic | |
Lanthanum(III) bromide, Ultra dry |
Beads | |
MFCD00016161 | |
XKUYOJZZLGFZTC-UHFFFAOYSA-K | |
lanthanum(3+) tribromide | |
378.62 | |
Ampule | |
(REO) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 236-896-7
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Ampouled under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only