missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) chloride, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

100.30 € - 1264.29 €

Chemical Identifiers

CAS 10099-58-8
Molecular Formula Cl3La
Molecular Weight (g/mol) 245.26
MDL Number MFCD00011068
InChI Key ICAKDTKJOYSXGC-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[La+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11368537
SDS View Documents
Thermo Scientific Alfa Aesar
035702.09
10 g
100.30 €
10g
Please to purchase this item. Need a web account? Register with us today!
11358537
SDS View Documents
Thermo Scientific Alfa Aesar
035702.18
50 g
328.64 €
50g
Please to purchase this item. Need a web account? Register with us today!
11415050
SDS View Documents
Thermo Scientific Alfa Aesar
035702.30
250 g
1264.29 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Lanthanum chloride solutions are used in various water treatment applications. It is also used in biochemical research. Doped with cerium, it is applied as a scintillator material. In organic synthesis, lanthanum trichloride functions as a mild lewis acid. This compound is used in gamma detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications

Chemical Identifiers

10099-58-8
245.26
ICAKDTKJOYSXGC-UHFFFAOYSA-K
[Cl-].[Cl-].[Cl-].[La+3]
Cl3La
MFCD00011068
lanthanum(3+) trichloride

Specifications

1,000°C
10099-58-8
Beads
MFCD00011068
14,5363
ICAKDTKJOYSXGC-UHFFFAOYSA-K
lanthanum(3+) trichloride
245.27
Odorless
Hygroscopic
(REO)
860°C
White
Cl3La
UN3077
Soluble in water.
[Cl-].[Cl-].[Cl-].[La+3]
245.26
99.9%
Ampule
3.842 g/mL
Lanthanum(III) chloride, Ultra dry
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H290-H317
Causes serious eye damage.
May be corrosive to metals.
May cause an allergic skin reaction.

P234-P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P390-P501c

H290-H317-H318

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III

missing translation for 'einecsNumber' : 233-237-5

missing translation for 'rtecsNumber' : OE4375000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.