Learn More
Lanthanum(III) oxalate decahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
82.01 € - 278.00 €
Chemical Identifiers
CAS | 537-03-1 |
---|---|
Molecular Formula | C6La2O12 |
Molecular Weight (g/mol) | 541.865 |
MDL Number | MFCD00150540 |
InChI Key | OXHNIMPTBAKYRS-UHFFFAOYSA-H |
Synonym | lanthanum oxalate, lanthanum iii oxalate, acmc-1bafu, lanthanum 3+ ; oxalate, dilanthanum 3+ trioxalate ion, dilanthanum 3+ trioxalate, lanthanum iii oxalate decahydrate, reacton, ethanedioic acid,lanthanum 3+ salt,hydrate 3:2:9, lanthanum, m-ethanedioato 2--ko1,ko2':ko1',ko2 bis ethanedioato 2--ko1,ko2 di |
PubChem CID | 164654 |
IUPAC Name | lanthanum(3+);oxalate |
SMILES | C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].[La+3].[La+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11392288
|
Thermo Scientific Alfa Aesar
013196.18 |
50 g |
82.01 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11382288
|
Thermo Scientific Alfa Aesar
013196.30 |
250 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
537-03-1 | |
541.865 | |
OXHNIMPTBAKYRS-UHFFFAOYSA-H | |
164654 | |
C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].[La+3].[La+3] |
C6La2O12 | |
MFCD00150540 | |
lanthanum oxalate, lanthanum iii oxalate, acmc-1bafu, lanthanum 3+ ; oxalate, dilanthanum 3+ trioxalate ion, dilanthanum 3+ trioxalate, lanthanum iii oxalate decahydrate, reacton, ethanedioic acid,lanthanum 3+ salt,hydrate 3:2:9, lanthanum, m-ethanedioato 2--ko1,ko2':ko1',ko2 bis ethanedioato 2--ko1,ko2 di | |
lanthanum(3+);oxalate |
Specifications
(decomposition) | |
White | |
C6La2O12 | |
MFCD00150540 | |
lanthanum oxalate, lanthanum iii oxalate, acmc-1bafu, lanthanum 3+ ; oxalate, dilanthanum 3+ trioxalate ion, dilanthanum 3+ trioxalate, lanthanum iii oxalate decahydrate, reacton, ethanedioic acid,lanthanum 3+ salt,hydrate 3:2:9, lanthanum, m-ethanedioato 2--ko1,ko2':ko1',ko2 bis ethanedioato 2--ko1,ko2 di | |
C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].[La+3].[La+3] | |
541.865 | |
722.02 (541.87 Anhydrous) | |
Reacton™ | |
(REO) |
537-03-1 | |
Powder | |
La2(C2O4)3·10H2O | |
UN3288 | |
OXHNIMPTBAKYRS-UHFFFAOYSA-H | |
lanthanum(3+);oxalate | |
164654 | |
99.99% | |
Hygroscopic | |
Lanthanum(III) oxalate decahydrate |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 208-656-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only