missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum matrix modifier solution, 99.999%, Honeywell™ Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
Description
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions

Specifications
Specifications
Quantity | 100 mL |
MDL Number | MFCD00011066 |
UN Number | UN1789 |
Percent Purity | 100.00% |
Density | 1.085g/mL at 25°C |
Chemical Name or Material | Lanthanum matrix modifier solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lanthanum matrix modifier solution, 99.999%, Honeywell™ Fluka™ >
Spot an opportunity for improvement?Share a Content Correction